Learn More
Bemegride, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Bemegride
Specifications
Specifications
| Chemical Name or Material | Bemegride |
| CAS | 64-65-3 |
| Analyte or Component Names | Bemegride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C8 H13 N O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Glutarimide, 3-ethyl-3-methyl- (8CI), 2,6-Dioxo-4-methyl-4-ethylpiperidine, 3-Ethyl-3-methylglutarimide, 3-Methyl-3-ethylglutarimide, 4-Ethyl-4-methyl-2,6-dioxopiperidine, 4-Ethyl-4-methyl-2,6-piperidinedione, 4-Methyl-4-ethyl-2,6-dioxopiperidine, Agipnon, Ahypnon, Antibarbi, Bemegrid, Bemegride, Etimid, Eukraton, Malysol, Megibal, Megimid, Megimide, Methetharimide, Mikedimide, NP 13, NSC 168667, NSC 58187, Zentraleptin, 2,6-Piperidinedione, 4-ethyl-4-methyl-, beta-Ethyl-beta-methylglutarimide, beta-Methyl-beta-ethylglutarimide |
| InChI Formula | InChI=1S/C8H13NO2/c1-3-8(2)4-6(10)9-7(11)5-8/h3-5H2,1-2H3,(H,9,10,11) |
| SMILES | CCC1(C)CC(=O)NC(=O)C1 |
| IUPAC Name | 4-ethyl-4-methylpiperidine-2,6-dione |
| Show More |