Learn More
Bemegride, MedChemExpress
Product Code. 16452266
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Product Code. 16452266
Brand: Medchem Express HYB1326
MedChemExpress Bemegride (3-Ethyl-3-methylglutarimide) is a central nervous system stimulant and antidote for barbiturate poisoning.
Bemegride (3-Ethyl-3-methylglutarimide) is a central nervous system stimulant and antidote for barbiturate poisoning.
Specifications
Chemical Name or Material Bemegride
CAS 64-65-3
Color White
Purity Grade Notes Research
Molecular Formula C8H13NO2
Quantity 500 mg
Synonym 3-Ethyl-3-methylglutarimide Bemegrid
Solubility Information DMSO : ≥ 32 mg/mL (206.20 mM)
SMILES O=C(CC(C)(CC)C1)NC1=O
Molecular Weight (g/mol) 155.19
Formula Weight 155.19
Percent Purity 99.92%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.