Learn More
Bemegride, LoGiCal
Product Code. 30462578
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. 30462578
Brand: LoGiCal LGCFOR1747.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Bemegride
CAS 64-65-3
Analyte or Component Names Bemegride
Shipping Condition Room Temperature
Molecular Formula C8 H13 N O2
Certifications/Compliance ISO 9001
Synonym Glutarimide, 3-ethyl-3-methyl- (8CI), 2,6-Dioxo-4-methyl-4-ethylpiperidine, 3-Ethyl-3-methylglutarimide, 3-Methyl-3-ethylglutarimide, 4-Ethyl-4-methyl-2,6-dioxopiperidine, 4-Ethyl-4-methyl-2,6-piperidinedione, 4-Methyl-4-ethyl-2,6-dioxopiperidine, Agipnon, Ahypnon, Antibarbi, Bemegrid, Bemegride, Etimid, Eukraton, Malysol, Megibal, Megimid, Megimide, Methetharimide, Mikedimide, NP 13, NSC 168667, NSC 58187, Zentraleptin, 2,6-Piperidinedione, 4-ethyl-4-methyl-, beta-Ethyl-beta-methylglutarimide, beta-Methyl-beta-ethylglutarimide
InChI Formula InChI=1S/C8H13NO2/c1-3-8(2)4-6(10)9-7(11)5-8/h3-5H2,1-2H3,(H,9,10,11)
Solution Type Neat
SMILES CCC1(C)CC(=O)NC(=O)C1
IUPAC Name 4-ethyl-4-methylpiperidine-2,6-dione
Industry Type Clinical Testing
Molecular Weight (g/mol) 155.19
Quantity 10 mg
Formula Weight 155.0946 g/mol