Learn More
Wilmad-LabGlass™ SP Scienceware™ 3mm O.D. Thin Walled Precision NMR Tube
Shop All SP Scienceware Products
Click to view available options
Length (Metric):
178 mm
203.2 mm
228.6 mm
Model:
305-PS-7-5
305-PS-8-5
305-PS-9-5
307-PP-7-5
307-PP-8-5
307-PP-9-5
327-PP-7-5
327-PP-8-5
327-PP-9-5
328-PP-7-5
328-PP-8-5
328-PP-9-5
335-PP-7-5
335-PP-8-5
335-PP-9-5
Field Strength:
200MHz
300MHz
400MHz
500MHz
600MHz
Unit Size:
Pack of 5
Description
To maximize SNR, Precision NMR Tubes have minimal paramagnetic impurities that would impact shimming. Tight I.D. and O.D. tolerance as small as 0.0065mm accommodates Wilmad inserts
Manufactured in a state-of-the-art ISO 9001:2008 USA facility using a unique precision shrinking and grinding process to shape the inner surface with maximized filling factor.
- Inner surface is resistant to strong acid and base at ambient temperature
- Can be operated safely at temperatures up to 230°C, and within a temperature step of 120°C
- Rated as hydrolytic class 1
- Made from borosilicate glass that meets the requirement of Type 1 Class A glass from ASTM E438
- 100% inspection with multiple NIST traceable gauges and optical surface defect checks
Specifications
Specifications
| Camber (Metric) | 25 μm |
| Concentricity (Metric) | 25μm |
| Length (Metric) | 178 mm |
| Material | Glass (Type 1, Class A) |
| Diameter (Metric) Outer | 2.9935±0.0065 mm |
| Diameter (Metric) Inner | 2.4195±0.0065 mm |
| Field Strength | 400MHz |
| Length (English) | 7 in. |
| Thickness (Metric) Wall | 0.29 mm |
| Model | 327-PP-7-5 |