Learn More
Bel-Art™ SP Scienceware™ Vikem™ Microscope Covers
Product Code. p-3207408 Shop All SP Scienceware Products
Click to view available options
Size:
L
Unit Size:
Each
Product Code. 15205428
Brand: Bel-Art™ F243030000
Only null left
Add to basket
Add to basket
Bel-Art™ SP Scienceware™ Vikem™ Microscope Covers are transparent vinyl covers that protect microscopes, balances and other instruments from dust and spills. Available in several sizes.
  • Cover slides over the instrument when not in use
  • Available in standard sizes
  • Sizes (bottom x top x height): Large size: 16 x 11 x 18 in. (41 x 28 x 46cm) and Extra-large size: 19 x 15 x 22 in. (46 x 38 x 59cm)
Specifications
Type Microscope Cover
Dimensions (L x W x H) 16 x 11 x 18 in. (41 x 28 x 46 cm)
Height (English) 18 in.
Height (Metric) 46 cm
Length (English) 16 in.
Length (Metric) 41 cm
Material Transparent Vinyl
Size L
Width (English) 11 in.
Width (Metric) 28 cm