Learn More
Bel-Art™ Touch Free™ Automatic Waste Cans
Product Code. p-2683129 Shop All SP Scienceware Products
Change view
Click to view available options
Dimensions (L x W x H):
14.4 x 10.5 x 22.5 in.
14.5 x 10.5 x 26.5 in.
14.625 x 10.5 x 25.75 in.
Capacity (English):
7.3 gal.
8.7 gal.
12 gal.
Color:
Gray
Red
Silver
Material:
ABS
Plastic
Stainless Steel
Unit Size:
Each
Product Code. Dimensions (L x W x H) Capacity (English) Color Material unitSize
12300629 14.4 x 10.5 x 22.5 in. 7.3 gal. Red ABS Each
11728414 14.625 x 10.5 x 25.75 in. 8.7 gal. Gray Plastic Each
11738414 14.5 x 10.5 x 26.5 in. 12 gal. Silver Stainless Steel Each
3 options
Product Code. 12300629
Brand: Bel-Art™ F132020022
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
Each unit is supplied with a brightly colored biohazard label and three other labels (recyclables, trash, and a blank label for individual applications).
Requires
Requires four D batteries (not included) which will provide approximately 4000 openings. All cans should be lined with disposable trash or biohazard bags (sold separately). Batteries and bags available on www.fishersci.com.
Automatically open and close without a touch, protecting users from waste receptacle contact.
  • When a hand or solid object is moved over the top, waste can detects motion, opens lid to accept waste, and then closes automatically
  • Fitted well helps to reduce odor and airborne contamination
  • Durable, dependable construction accommodates Bel-Art™ liners, biohazard liners or commercially available trash bags
  • ON-OFF switch deactivates AUTO-OPEN/CLOSE function
  • 2 Gallon (Cat. No. 11-394-451)
  • Ideal for benchtop use
  • Removable rigid autoclavable polypropylene liner will not leak or rust
  • Body and lid made from easy-to-clean, molded ABS plastic
  • White pail with white top
  • Line with 14 x 19 in. biohazard disposal bags or suitable size standard trash bags
7.3 Gallon (Cat. No. 11-394-452)
  • Pail made from polypropylene with an ABS plastic top
  • Line with 24 x 30 in., 24 x 36 in. or 25 x 35 in. biohazard disposal bags or suitable size standard trash bags
  • Gray pail with gray top
  • Line with 24 x 30 in., 24 x 36 in. or 25 x 35 in. biohazard disposal bags or suitable size standard trash bags
  • 7.3 Gallon (Cat. No. 03-409-55)
  • Pail made from polypropylene with an ABS plastic top
  • Line with 24 x 30 in., 24 x 36 in. or 25 x 35 in. biohazard disposal bags or suitable size standard trash bags
  • Red pail with red top
  • Line with 24 x 30 in., 24 x 36 in. or 25 x 35 in. biohazard disposal bags or suitable size standard trash bags
  • 12 Gallon (Cat. No. 11-394-453)
  • Small footprint enables a fit into tight spaces
  • Stainless-steel pail with black ABS plastic top
  • Line with 24 x 30 in., 24 x 36 in. or 25 x 35 in. biohazard disposal bags or suitable size standard trash bags
  • Prior to use, cans should be appropriately labeled with one of the included labels to indicate the type of waste to be placed inside the can.
Specifications
Capacity (Metric) 27.6 L
Capacity (English) 7.3 gal.
Color Red
Dimensions (L x W x H) 14.4 x 10.5 x 22.5 in.
Height (Metric) 57.1 cm
Length (Metric) 36.8 cm
Width (Metric) 26.7 cm
Autoclavable Autoclavable
Includes Brightly colored biohazard label and three other labels (recyclables, trash, and a blank label for individual applications)
Width (English) 10.5 in.
Length (English) 14.4 in.
Height (English) 22.5 in.
Material ABS
Product Type Automatic Waste Can