Learn More
Bel Art™ Test Tube Baskets with Lids
Product Code. 10114194
Change view
Click to view available options
Dimensions (L x W x H):
5 x 5.18 x 4 in.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Unit Size
10114194 5 x 5.18 x 4 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10114194 Supplier Bel Art™ Supplier No. F187370010
  • ONLINE EXCLUSIVE PRICE
    525.00 NOK
    378.39 NOK / Pack of 1
    Save 146.61 NOK
    27% Off
Estimated Shipment: 05-01-2027
Perfect for dipping, transporting and storing test tubes.
  • Polypropylene
  • Can be used for transporting small lab animals
Specifications
Autoclavable Not Autoclavable
Color White
Holds Test Tubes
Material Polypropylene
Shape Square
Stackable Yes
Sterility Non-sterile
For Use With (Equipment) Test Tubes
Size 5 x 4 x 4 in.
Dimensions (L x W x H) 5 x 5.18 x 4 in.
Length (Metric) 12.7 cm
Length (English) 5 in.
Width (English) 5.18 in.
Height (English) 4 in.
Product Type Basket for Test Tubes
Diameter (Metric) Inner 101.6 mm
Height (Metric) 10.1 cm
Array 1 x 1
Width (Metric) 13.1 cm