Learn More
Bel-Art™ T Connectors
Product Code. p-3273411 Shop All SP Scienceware Products
Click to view available options
For Use With:
9.5mm
For 4.7mm Tubing I.D.
Unit Size:
Pack of 12
Product Code. 11714935
Brand: Bel-Art™ F196060000
Only null left
Add to basket
Add to basket
Connectors have multiple barbs per arm providing additional contact between the fitting and the tubing for extra gripping power
  • Ensures a tight, secure connection between lengths of tubing having the same inner diameter (I.D.)
  • Chemical-resistant polypropylene for use with a wide range of liquids or gases
  • FDA-grade for direct contact with food, drugs, and cosmetics
  • Autoclavable
Specifications
Certifications/Compliance FDA
Autoclavable Autoclavable
Material Polypropylene
For Use With 9.5mm
Shape T
Color Natural
Product Type Tubing Connector