Description
- Protect against dangerous tears and leaks
- Orange-red bags have standard biohazard warning symbol and precautionary procedures in four languages: English, French, German, and Spanish
- Extra-heavy-duty bags are 2 mil-thick blended polymer
- Autoclavable to 121°C (250°F)
- Sterilization indicator patch darkens when exposed to steam sterilization
Specifications
Specifications
| Capacity (English) | 25 gal. |
| Color | Red |
| Dimensions (L x W) | 31 x 38 in. (79 x 96 cm) |
| Autoclavable | Autoclavable |
| Includes | Standard biohazard warning symbol and precautionary procedures in four languages: English, French, German and Spanish |
| For Use With (Application) | Extra-strength, high-impact, high-temperature bags reduce autoclave time |
| Thickness | 2 mil |
| Material | Polymer |
| Product Type | Biohazard Disposal Bag |