Learn More
Bel-Art™ Sterileware™ Economy Sampling Scoops
Product Code. 11332174 Shop All SP Scienceware Products
Change view
Click to view available options
Capacity (Metric):
63 mL
83 mL
125 mL
250 mL
Unit Size:
Pack of 50
Product Code. Capacity (Metric) unitSize
11332174 83 mL Pack of 50
11322174 63 mL Pack of 50
11342174 125 mL Pack of 50
11352174 250 mL Pack of 50
4 options
Product Code. 11332174
Brand: Bel-Art™ F369330002
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
An inexpensive and accurate measure for dry or liquid sample collection.
  • Capacity is listed on each white scoop
  • Leveling the sample to the flat top edge ensures a repeatable and consistent volume measure
  • Flat bottom shape and light weight allows for use as a weighing boat
  • One-piece scoops are made from durable, FDA grade, high-impact polystyrene plastic
  • Sterile are individually sealed and sterilized in a polyethylene bag ready for one time use and disposal, lot stamped for accurate lot tracing and shipped with a Certificate of Processing for Sterilization
Specifications
Quantity 50/Pk.
Sterility Sterile
Autoclavable Not Autoclavable
Shape Flat Bottom
Capacity (English) 2.7 oz.
Capacity (Metric) 83 mL
Color White
Certifications/Compliance FDA
Disposable Single-use
Product Type Scoop
For Use With (Application) Provide accurate measure for dry or liquid measures
Length (English) 59/16 in.
Size 10.3 x 4.9 x 2.9 cm (LxWxH) Bowl
Width (English) 1¹5/16 in.
Width (Metric) 4.9 cm
Includes Certificate of processing for sterilization
Length (English) Handle 0.50 in.
Length (Metric) Handle 3.8 cm
Material High-impact Polystyrene (FDA Grade)
Length (Metric) 14.1 cm
Show More Show Less
Sterilized products are not returnable.