Learn More
Bel-Art™ Sterileware™ Disposable Sterile Sampler Knife
Product Code. 11336944
Change view
Click to view available options
Length (English):
8.25 in.
Unit Size:
Pack of 200
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English) Unit Size
11336944 8.25 in. Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11336944 Supplier Bel-Art™ Supplier No. H369650000
  • ONLINE EXCLUSIVE PRICE
    4175.00 NOK
    3641.77 NOK / Pack of 200
    Save 533.23 NOK
    12% Off
 Restricted Item
Estimated Shipment: 14-12-2026
Polystyrene sampler knife featuring a blade that is serrated on one edge and smooth on the other
  • The serrated edge is capable of cutting seafood, meat, poultry, vegetables and other fibrous materials
  • Individually wrapped, sterilized and packed in a polyethylene (PE) plastic bag for one-time use and disposal
Specifications
Product Type Sampling Knife
Material Polystyrene
Includes Certificate of Processing for Sterilization
Disposable Single-use
Sterility Sterile
For Use With (Application) Food sampling like Meats, Vegetables, Seafood and other Fibrous and Semi Solid Materials
Length (English) 8.25 in.
Length (Metric) 20.9 cm