Learn More
Bel-Art™ Spray Pump Bottles
Product Code. p-3273679 Shop All SP Scienceware Products
Click to view available options
Capacity (Metric):
250 mL
Unit Size:
Pack of 12
Product Code. 13157256
Brand: Bel-Art™ F116330000
Only null left
Add to basket
Add to basket
Low-density polyethylene plunger style bottle delivers a fine mist spray.
  • Includes clear cap to help prevent evaporation and accidental leaks
  • 12/Pk.
Specifications
Capacity 8 oz., 250 mL
Capacity (Metric) 250 mL
Capacity (English) 8 oz.
Color Translucent, White
Material LDPE
Quantity 12/Pk.
Sterility Non-sterile
Closure Included Yes
Disposable Reusable
Graduated No
Mouth Narrow, Threaded
Shape Cylindrical
Height (Metric) 171.5 mm
Product Type Spray Bottle
Width (English) 2.33 in.
Length (English) 6.75 in.
Height (English) 6.75 in.
Chemical Label Blank
Closure Material Polystyrene
Closure Size 18 mm
Diameter (Metric) 59 mm
Closure Type Threaded