Description
- Small removable pivot ring in center adds to versatility
- Pivot ring minimizes contact area of bar to vessel, reduces friction and lessens marring of plastic containers
- Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE low friction coating
- Durable and inert
- Autoclave safe
Certifications
Manufactured in a registered ISO 9001:2008, as verified by SGS certification
Specifications
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | FDA, USP Class VI, ISO 9001:2008 |
| Color | White |
| Diameter (English) | 0.75 in. |
| Length (English) | 6 in. |
| Coating | PTFE, FDA-grade, USP Class VI |
| Diameter (Metric) | 19 mm |
| Length (Metric) | 152.4 mm |
| Material | Alnico Magnet |
| Quantity | 50 |
| Show More |