Learn More
Bel Art™ Poxygrid™ Baskets
Product Code. 11763207
Change view
Click to view available options
Dimensions (L x W x H):
24.75 x 14 x 6 in.
11 x 6 x 3.5 in.
14 x 11 x 3.5 in.
20 x 11 x 3.5 in.
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) unitSize
11763207 24.75 x 14 x 6 in. Each
11773207 11 x 6 x 3.5 in. Each
11793207 20 x 11 x 3.5 in. Each
11783207 14 x 11 x 3.5 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11763207 Supplier Bel Art™ Supplier No. H167590000
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
Epoxy-coated steel wire construction holds up under severe laboratory conditions
Epoxy-coated steel wire construction holds up under severe laboratory conditions
  • Resist rust, corrosion, solvents, acids and alkalis
  • Convenient for transporting and batch-dipping glassware
  • Steam autoclavable at 121°C (250°F)
Specifications
Autoclavable Autoclavable
Color Blue
Holds Labware
Material Epoxy-coated Steel
Shape Rectangular
Stackable No
Sterility Non-sterile
Size 630 x 360 x 150 mm
Dimensions (L x W x H) 24.75 x 14 x 6 in.
Length (Metric) 63 cm
Length (English) 24.75 in.
Width (English) 14 in.
Height (English) 6 in.
Includes Basket Only
Product Type Wire Basket
Height (Metric) 15.2 cm
Array 1 x 1
Width (Metric) 36 cm