Learn More
Bel-Art™ Nylon Screw Compressor Clamp
Product Code. 11739695 Shop All SP Scienceware Products
Change view
Click to view available options
Color:
Brown
Unit Size:
Pack of 12
Product Code. Color unitSize
11739695 Brown Pack of 12
1 options
Product Code. 11739695
Brand: Bel-Art™ F182250000
Only null left
Add to basket
Add to basket
Tubing clamp with angled jaws prevents tube from slipping out
Tubing clamp with angled jaws prevents tube from slipping out
  • Open-jaw nylon tubing clamp fits any tubing up to 1/4 in. O.D. (6.4mm)
  • Thumbscrew clamp applies even force
Specifications
Quantity 12 Pk.
Type Screw Compressor Clamp
Material Nylon
For Use With (Equipment) 0.25 in. O.D. Tube
Length (English) 2.31 in.
Length (English) Opening 0 to 0.25 in.
Length (Metric) 5.867 cm
Length (Metric) Opening 0 to 0.63 cm
Color Brown
Jaw Type Angled
Diameter (Metric) Inside 6.4 mm