Learn More
Bel-Art™ Mini Sampler Spoons
Product Code. 15590102 Shop All SP Scienceware Products
Change view
Click to view available options
Capacity (Metric):
0.05 mL
0.1 mL
0.25 mL
0.5 mL
1 mL
Unit Size:
Each
Pack of 25
Product Code. Capacity (Metric) unitSize
15590102 0.5 mL Pack of 25
15570102 0.1 mL Pack of 25
12870953 0.05 mL Each
15580102 0.25 mL Pack of 25
15500112 1 mL Pack of 25
5 options
Product Code. 15590102
Brand: Bel-Art™ F367210050
Only null left
Add to basket
Add to basket
Avoid cross contamination by using a different spoon for each sample
  • These disposable mini measures are designed for retrieving small, specific volume samples of powders or similar materials
  • Made from white high-impact polystyrene plastic
  • All sterile products are lot stamped for accurate tracing
Specifications
Sterility Sterile
Autoclavable Not Autoclavable
Capacity (Metric) 0.5 mL
Color White
Disposable Single-use
Product Type Spoon
For Use With (Application) Designed for retrieving small, specific volume samples of powders or similar materials
Includes Rounded bowl and thin handle neck
Material Polystyrene