Learn More
Bel-Art™ Mini Sampler Spoons
Product Code. 15500112 Shop All SP Scienceware Products
Click to view available options
Capacity (Metric):
0.05 mL
0.1 mL
0.25 mL
0.5 mL
1 mL
Unit Size:
Each
Pack of 25
Product Code. 15500112
Brand: Bel-Art™ F367210100
Only null left
Add to basket
Add to basket
Avoid cross contamination by using a different spoon for each sample
  • These disposable mini measures are designed for retrieving small, specific volume samples of powders or similar materials
  • Made from white high-impact polystyrene plastic
  • All sterile products are lot stamped for accurate tracing
Specifications
Sterility Sterile
Autoclavable Not Autoclavable
Capacity (Metric) 1 mL
Color White
Disposable Single-use
Product Type Spoon
For Use With (Application) Designed for retrieving small, specific volume samples of powders or similar materials
Includes Rounded bowl and thin handle neck
Material Polystyrene