Learn More
Bel-Art™ Mercury Collector
Shop All SP Scienceware Products
Click to view available options
For Use With (Application):
Mercury spill clean-up
Mercury spill clean-up replacement pad
Unit Size:
Each
Pack of 12
Description
Captures spilled mercury beads quickly and conveniently. Bel-Art™ Mercury Collector holds 125 ml of mercury.
- Lid features absorbent foam pad on interior surface
- Replacement foam pad available
- Dispose of mercury by approved disposal method
Specifications
Specifications
| Color | White |
| Capacity (Metric) | 125 mL |
| Material | Polyethylene |
| Type | Replacement Pad |
| For Use With (Application) | Mercury spill clean-up replacement pad |
| Includes | Jar, perforated plate, lid with attached foam pad |
| Height (English) | 1.88 in. |
| Height (Metric) | 4.77 cm |