Learn More
Bel Art™ Lab Companion Cabinet Style Vacuum Desiccator, Clear
Product Code. 15369807
Change view
Click to view available options
Volume (Metric):
11 L
23 L
35 L
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15369807 35 L Each
17492883 11 L Each
15605439 23 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15369807 Supplier Bel Art™ Supplier No. F424004011
  • ONLINE EXCLUSIVE PRICE
    24795.00 NOK
    19392.55 NOK / Pack of 1
    Save 5402.45 NOK
    21% Off
Estimated Shipment: 25-11-2026
Includes
Desiccant tray (desiccants sold separately)
Clear desiccator provides vacuum sustainment in a space maximizing cube.
  • Able to maintain 29 in.-Hg vacuum for over 72 hours at room temperature, they stay gastight allowing reliable storage or experiments over extended periods
  • Viton™ 3-way valve offers consistent and uniform vacuum draw, vacuum release, or gas exchange
  • Transparent polycarbonate construction is durable, easy-to-clean, chemical-resistant, and allows quick visualization of the desiccator contents
  • Amber tinted UV blocking models block 100% of UVA, B, and C light to minimize damages or discoloration of light-sensitive samples
  • Use with any vacuum with free air capacity of 1.5 cubic feet per minute (CFM) ft or greater
  • Single piece molded design (except the door) enables outstanding vacuum capability with virtually no leakage and exceptional durability
  • Cabinet style provides maximum useable space
  • Hinged door allows easy access; built in locking tabs secure the door when not under vacuum
  • Analog vacuum gauge in front door allows vacuum level checking at a glance
  • Perforated shelves can be repositioned to accommodate various storage needs (additional shelves sold separately)
  • Built-in ports permit attachment of up to 3 additional valves (sold separately)
  • Stackable space-saving design allows efficient use of work space
  • Eight models to choose from; 11, 23, 35, and 45 liter internal volume in clear or UV blocking amber
Specifications
Color Clear
Material Polycarbonate
Product Type Vacuum Desiccator
Door Type Hinged
Includes Desiccant tray, 3 perforated shelves, built-in ports, door-mounted analog vacuum gauge
Max. Shelf Load 5 kg
No. of Shelves 3/5
Style Cabinet
Volume (English) 1.2 cu. ft.
Volume (Metric) 35 L
Dimensions (D x W x H) Interior 14.75 x 14 x 13.62 in. (37.46 x 35.56 x 34.59 cm)
Width (English) 16.5 in.
Width (English) Door 11.6 in.
Width (English) Interior 14 in.
Width (Metric) 42 cm
Width (Metric) Door 29.5 cm
Width (Metric) Interior 35.56 cm
Height (Metric) 38.1 cm
Height (English) 15 in.
Height (English) Door Opening 13.6 in.
Height (English) Interior 13.62 in.
Height (Metric) Door Opening 34.5 cm
Height (Metric) Interior 34.59 cm
Depth (English) 15.62 in.
Depth (English) Interior 14.75 in.
Depth (Metric) 39.7 cm
Depth (Metric) Interior 37.46 cm
Diameter (English) Outer 15.62 in.
Diameter (Metric) Inner 35.6 cm
Diameter (Metric) Outer 39.67 cm
Weight (English) 23.7 lb.
Weight (Metric) 10.8 kg