Learn More
Bel Art™ Lab-Aire™ II Glassware Drying Racks
Product Code. 11882801 Shop All SP Scienceware Products
114, Benchtop Rack (3-tier), 22.4 in., 10 in., Each
Click to view available options
Positions:
19
38
57
76
114
Product Type:
Benchtop Rack (1-tier)
Benchtop Rack (2-tier)
Benchtop Rack (3-tier)
Benchtop/Wall-mount Rack (1-tier)
Wall-mount Rack (2-tier)
Wall-mount Rack (3-tier)
Height (English):
8.4 in.
9.4 in.
15.4 in.
16.4 in.
22.4 in.
23.4 in.
Depth (English):
5 in.
7 in.
10 in.
Unit Size:
Each
Product Code. 11882801
Brand: Bel Art™ F189330023
Only undefined left
Only undefined left
Add to basket
Add to basket
Glassware and plasticware drying racks for walls or benchtops
Glassware and plasticware drying racks for walls or benchtops
  • Dry and store delicate labware including beakers, flasks, bottles, test tubes and cylinders on these space-saving drying racks
  • Single-sided benchtop models fit flat against a wall
  • Double-sided benchtop models provide double the capacity (up to 114 pegs) with a slightly larger footprint
  • Three sizes and lengths of V-channel pegs can be repositioned to hold a variety of labware
  • Rugged ABS plastic body and pegs wipe clean and won't corrode
  • Wall-mount models have mounting points that fit standard pegboards
  • Pegs available separately to reconfigure rack for specific drying requirements
Specifications
Includes Pegs
Product Type Benchtop Rack (3-tier)
Array 114 x 1
Autoclavable Not Autoclavable
Color Cream, Teal
Holds Lab Glassware, Lab Plasticware
Material ABS
Positions 114
Shape Rectangular
Shelves None
Stackable No
Sterility Non-sterile
For Use With (Equipment) Lab Glassware
Size 425 x 254 x 569 mm
Length (Metric) 577 mm
Width (Metric) 37.5 cm
Width (English) 14.75 in.
Height (Metric) 57 cm
Height (English) 22.4 in.
Depth (English) 10 in.
Depth (Metric) 25.4 cm
Product Title
Bel Art™ Lab-Aire™ II Glassware Drying Racks >