Learn More
Bel-Art™ Glove Box Economy Sleeve Gloves
Shop All SP Scienceware Products
Click to view available options
Size:
9
10
Unit Size:
Pack of 2
Description
- Single piece glove with sleeve
- For glove boxes with 8 in. glove ports
- Requires clamping rings (19-821-983) to hold gloves in place
- Sold in pairs
Specifications
Specifications
| Color | Black |
| Hand | Specific |
| Latex Free | No |
| Material | Bonded Neoprene |
| Product Type | Glove Box Gloves |
| Size | 10 |