Learn More
Bel-Art™ Glascribe™ Pen
Product Code. 11327354
Change view
Click to view available options
Quantity:
Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11327354 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11327354 Supplier Bel-Art™ Supplier No. H441500000
  • ONLINE EXCLUSIVE PRICE
    417.00 NOK
    352.05 NOK / Pack of 1
    Save 64.95 NOK
    15% Off
View all promotions
Estimated Shipment: 04-12-2026
Permanently marks laboratory glassware, ceramics and plastics
  • Tungsten carbide tip (Mohs Hardness = 9) easily inscribes identifying marks on labware
  • Retractible tip operates with pen style push button
  • Clips to a pocket, ready for use
Specifications
Tip Style Retractable
Product Type Scribe Pen
Color Black
Tip Material Tungsten Carbide
Includes Pocket clip
For Use With (Application) Permanently marks laboratory glassware, ceramics and plastics
Quantity Each