Learn More
Bel-Art™ Flea Micro Spinbar™ Magnetic Stirring Bars
Product Code. 11398703 Shop All SP Scienceware Products
Click to view available options
Length (Metric):
7 mm
8 mm
10 mm
Diameter (Metric):
1.5 mm
2 mm
3 mm
Shape:
Cylinder
Octagon
Coating:
PTFE
PTFE, FDA-grade, USP Class VI
Unit Size:
Each
Product Code. 11398703
Brand: Bel-Art™ F371190007
Only null left
Add to basket
Add to basket
Particularly useful for environmental testing and life science applications in which small sample volumes need to be prepared and evaluated
  • Small size bar for stirring in vessels such as vials, micro test tubes and centrifuge tubes
  • Alnico magnet encapsulated in FDA Grade
  • USP Class VI PTFE low friction coating
  • Durable and inert
  • Autoclave safe
  • Particularly useful for environmental testing and life science applications in which small sample volumes need to be prepared and evaluated
Certifications
Manufactured in a registered ISO 9001:2008, as verified by SGS certification
Specifications
Autoclavable Autoclavable
Certifications/Compliance FDA, USP Class VI, ISO 9001:2008
Color White
Diameter (English) 0.07 in.
Length (English) 0.27 in.
Coating PTFE, FDA-grade, USP Class VI
Diameter (Metric) 2 mm
Length (Metric) 7 mm
Material Alnico Magnet
Quantity 50
Shape Octagon
Type Magnetic Stir Bar, Micro FLEA