Learn More
Bel-Art™ SP Scienceware™ Double Spinfin™ Magnetic Stirring Bars
To agitate liquids in round bottom vessels where other stirring bars cannot be used.
Brand:  Bel-Art™ F37126-0001
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11308723
  • 260.00 NOK / Each
Estimated Shipment: 12-07-2024
Description
  • Made from PTFE
  • Fins on top and bottom ensure complete agitation
  • Does not wobble, maintains position in vessel
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE low friction coating
  • Durable and inert
  • Autoclavable
Specifications
Specifications
25.4 mm
19.1mm
PTFE
1 in.
0.75 in.