Learn More
Bel-Art™ Cover Glass Forceps
Product Code. p-3205717 Shop All SP Scienceware Products
Click to view available options
Length (Metric):
102 mm
114 mm
Tip Style:
Bent
Straight
Unit Size:
Pack of 2
Product Code. 12822031
Brand: Bel-Art™ H379430000
Only null left
Add to basket
Add to basket
Tips shaped to easily lift cover glass. Bel-Art™ Cover Glass Forceps are fully coated with brown Teflon™ FEP and comply with FDA regulations concerning direct food contact.
  • Brown PTFE FEP coating guards against scratches
  • Wide, bent tip style is perfect for lifting cover glasses
Specifications
Tip Style Straight
Length (English) 4.5 in.
Material PTFE
Length (Metric) 114 mm
Product Type Forceps
Finish Bbrown PTFE-FEP coating
For Use With (Application) Lifting cover glasses
Type FEP Coated Cover Glass Forceps
Style Slide Holding