Learn More
Bel-Art™ Connector Assortment
Product Code. 11318691
Change view
Click to view available options
Shape:
Straight
Unit Size:
Pack of 22
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Shape Unit Size
11318691 Straight Pack of 22
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11318691 Supplier Bel-Art™ Supplier No. H195700000
  • ONLINE EXCLUSIVE PRICE
    877.00 NOK
    744.87 NOK / Pack of 22
    Save 132.13 NOK
    15% Off
View all promotions
Estimated Shipment: 22-12-2026
Collection of polypropylene straight and straight-stepped tubing connectors
This assortment of 22 stepped and straight tubing connectors comes in handy around the laboratory, and contains fittings compatible with tubing from 3.2 to 12.7mm (0.125 to 0.5 in.).
  • Chemical-resistant polypropylene for use with wide range of liquids or gases
  • Includes 12 Stepped Tubing Connectors and 10 Straight Tubing Connectors
  • FDA-grade polypropylene for direct contact with food, drugs, and cosmetics
  • Autoclavable
Specifications
Autoclavable Autoclavable
Material Polypropylene
Shape Straight
Color White
Product Type Tube Connector