Learn More
Bel-Art™ Clear Graduated Cylinders
Product Code. p-4754974 Shop All SP Scienceware Products
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Graduations:
0.1 mL
0.5 mL
1 mL
2 mL
5 mL
10 mL
20 mL
Unit Size:
Each
Product Code. 30193476
Brand: Bel-Art™ F286910000
Only null left
Add to basket
Add to basket
No concave meniscus, makes the raised numeral, single scale, graduations simple to interpret.
  • Made from clear TPX
  • Shatter-proof
  • Wide octagonal base prevents tipping and rolling
  • Chemical and heat resistant to 177°C (350°F)
  • Steam autoclavable at 121°C (250°F)
Specifications
Capacity (Metric) 25 mL
Material Plastic
Tolerance Chemical, Heat Resistant
Diameter (English) 0.7 in.
Scale Single
Graduations 0.5 mL
Autoclavable Autoclavable
Subdivision 0.5 mL
Shape Round
Base Type Octagonal
Height (English) 7.5 in.
Height (Metric) 190.5 mm
Type Graduated Cylinder