Learn More
Bel-Art™ Clavies™ High-Temperature Autoclave Bags
Product Code. p-3207300 Shop All SP Scienceware Products
Change view
Click to view available options
Dimensions (L x W):
10 x 10 in.
10 x 8 in. (25.4 x 20cm)
12 x 10 in. (30.48 x 25.4cm)
12 x 8 in. (30 x 20 cm)
15 x 10 in. (38.1 x 25.4cm)
15 x 8 in. (38.1 x 20cm)
24 x 12 in. (61 x 30 cm)
30 x 24 in. (76 x 61 cm)
36 x 24 in. (91 x 61 cm)
Unit Size:
Pack of 100
Product Code. Dimensions (L x W) unitSize
11774286 10 x 10 in. Pack of 100
13293529 15 x 8 in. (38.1 x 20cm) Pack of 100
15421690 12 x 10 in. (30.48 x 25.4cm) Pack of 100
15520513 15 x 10 in. (38.1 x 25.4cm) Pack of 100
10768895 36 x 24 in. (91 x 61 cm) Pack of 100
10799465 30 x 24 in. (76 x 61 cm) Pack of 100
10490955 24 x 12 in. (61 x 30 cm) Pack of 100
13273529 12 x 8 in. (30 x 20 cm) Pack of 100
12695457 10 x 8 in. (25.4 x 20cm) Pack of 100
9 options
Product Code. 11774286
Brand: Bel-Art™ H131821010
Only null left
Add to basket
Add to basket
Includes
Bag closures and holders sold separately.
Withstand high temperatures to reduce autoclave time.
Withstand high temperatures to reduce autoclave time
  • Suitable for discarding Petri dishes, membrane filters, multi-well culture plates, pipets and more
  • Rugged, transparent, 0.05 mm (2 mil) thick polypropylene
  • Steam autoclavable at 275°F (135°C)
Specifications
Autoclavable Autoclavable
Capacity (English) 0.85 gal.
Certifications/Compliance Dart Impact: 1000g per ASTM D1709-01. Tear Resistance: ASTM D1922-00A, MD 200g.
Closure Type None
Color Clear
Disposable Single-use
Material Polypropylene
Sterility Non-sterile
Thickness (Metric) 0.050 mm
Thickness (English) 2 mil
Dimensions (L x W) 10 x 10 in.
Length (English) 10 in.
Length (Metric) 25.4 cm
Width (English) 10 in.
Width (Metric) 25.4 cm
Product Type Autoclave Bags