Learn More
Bel-Art™ Captor™ Pen Holder
Product Code. 11737774
Change view
Click to view available options
Quantity:
3 Pk.
Unit Size:
Pack of 3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11737774 3 Pk. Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11737774 Supplier Bel-Art™ Supplier No. F133730000
Stock Up Your Lab
Use promo code 28007
View all promotions
Estimated Shipment: 06-10-2026
Add to basket
Add to basket
Cap and uncap pen with one hand
  • Avoid fumbling with caps and risking contamination of samples
  • Fits Sharpie™ markers or similar pens with a pocket clip
  • Attaches with press-fit Dual Lock™ reclosable fastener to any flat surface, including incubator, freezer, centrifuge, fume hood or benchtop
  • Black polycarbonate
  • 1L × 1W × 2 in.H (2.5 × 2.5 × 5.1cm)
Specifications
Tip Style Felt Tip
Product Type Pen Holder
Color Black
Material Polycarbonate
For Use With (Application) Fits Sharpie™ markers or similar pens with a pocket clip
For Use With (Equipment) Attaches with press-fit Dual Lock reclosable fastener
Quantity 3 Pk.
Height (English) 2 in.
Length (English) 1 in.
Width (English) 1 in.