Learn More
Bel-Art™ Glove Box Accessory, Iris Port Entry
Product Code. 10646103
Change view
Click to view available options
Material:
Silicone Latex Rubber
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
10646103 Silicone Latex Rubber Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10646103 Supplier Bel-Art™ Supplier No. H500250020
  • ONLINE EXCLUSIVE PRICE
    10950.00 NOK
    9553.28 NOK / Pack of 2
    Save 1396.72 NOK
    12% Off
 Restricted Item
Estimated Shipment: 12-01-2027
Bare Hand Entry (Iris) Port allows bare hnd entry and operation.
  • Fits 7.75 in. I.D. (19.7 cm) glove ports
  • Made of ten layers of soft, conforming rubber
  • Easy access and withdrawal, plus excellent sealing
Order Info
One pair per pack.
Specifications
Product Type Glove Box Port Cover
Material Silicone Latex Rubber
For Use With (Equipment) 19.7cm I.D. glove ports