Learn More
Bel-Art™ SP Scienceware™ Barbed Fittings with Reducing Connector
Description
- Chemical-resistant polypropylene plastic for use with wide range of liquids or gases
- FDA-grade for direct contact with food, drugs, and cosmetics
- Autoclavable
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Polypropylene |
| For Use With | 4.7 to 6mm |
| Tubing Size | For 3/16 in. to 1/4 in. tubing (4.7 to 6mm) |
| Includes | Reducing connector |
| Color | Clear |
| Product Type | Barbed Fitting |