Learn More
Bel-Art™ SP Scienceware™ Barbed Fittings with Reducing Connector
Product Code. 15102335 Shop All SP Scienceware Products
Click to view available options
For Use With:
4.7 to 6mm
4.7 to 9.5mm
6 to 12.7mm
9.5 to 12.7mm
Fits 9 x 6mm Tubing I.D.
Unit Size:
Pack of 12
Product Code. 15102335
Brand: Bel-Art™ 0645860
Only null left
Add to basket
Add to basket
These fittings ensure a tight, secure connection between tubing having different inner diameters (I.D.)
  • Chemical-resistant polypropylene plastic for use with wide range of liquids or gases
  • FDA-grade for direct contact with food, drugs, and cosmetics
  • Autoclavable
Specifications
Autoclavable Autoclavable
Material Polypropylene
For Use With 9.5 to 12.7mm
Tubing Size For 3/8 in. to 1/2 in. tubing (9.5 to 12.7mm)
Includes Reducing connector
Color Clear
Product Type Barbed Fitting