Learn More
Bel-Art™ Bag Twist Tie Cord
Product Code. 11707035
Change view
Click to view available options
Length (Metric):
457.2 m
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
11707035 457.2 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11707035 Supplier Bel-Art™ Supplier No. H131900000
  • ONLINE EXCLUSIVE PRICE
    1045.00 NOK
    889.04 NOK / Pack of 1
    Save 155.96 NOK
    14% Off
Estimated Shipment: 27-11-2026
Strong twist tie bag closures easier to use than string or rubber bands
  • Cut to optimum length and twist together for best closure performance
  • Use to bundle hard-to-handle items like rods, wires or tubes
  • Made of soft, copper-tinned, annealed iron wire sealed in extruded virgin vinyl
  • Autoclavable 121°C for 30 minutes
  • 1500 ft. per roll
Specifications
Type Ties
Material Nylon
Length (Metric) 457.2 m
Length (English) 1500 ft
For Use With (Application) Use to close bags or to package rods, wires and tubes