Description
- Acrylic cabinet with removable, tray-like shelves with perforated bottoms and slide in/out on pre-set rails
- Door opens from right to left and secured with stainless steel latch
- Available with three shelves
- Comes in clear color and bronze color for light-sensitive materials
Specifications
Specifications
| Color | Bronze |
| Cabinet Style | Front-loading |
| Material | Acrylic |
| Product Type | Desiccator |
| Door Style | Gasketed |
| For Use With (Application) | Used in refrigerators/at room temperature for light-sensitive materials |
| Holds | 32 Cans |
| Includes | Tray-like 3 shelves with perforated bottoms and slide in/out on pre-set rails, stainless steel twist locking latch for door |
| No. of Shelves | 3 removable tray-like shelves |
| Volume (English) | 0.82 cu. ft. |
| Show More |