Description
- Bright red color reduces chance of scoop being left behind in a sample; can serve as code to avoid cross contamination; can provide contrasting background for light colored samples
- High handle location keeps hand further from sample
- Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
- High impact polystyrene (HIPS) scoops heat sealed in individual polyethylene bags
- Gamma irradiated sterile (SAL 10^-6)
- Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization
Specifications
Specifications
| Quantity | 25/Pk. |
| Autoclavable | Not Autoclavable |
| Capacity (Metric) | 1000 mL |
| Color | Red |
| Product Type | Scoop |
| Dimensions (L x W) | 17.5 x 15 x 9.6 cm |
| Length (Metric) Handle | 14.8 cm |
| Material | High-impact Polystyrene (FDA Grade) |