missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beclin 2 Antibody [Janelia Fluor™ 669], R&D Systems™
Description
Beclin 2 Polyclonal antibody specifically detects Beclin 2 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Beclin 2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 669 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | BECN1 L1, BECN1L1, BECN1P1, BECN1P1 beclin 1, autophagy related, pseudogene 1 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide made to an N-terminal region (within residues 1-100) of the human Beclin 2 protein. [Uniprot # A8MW95] |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?