Learn More
Beclin 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Product Code. 16828293 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16828293 20 μL 20µL
16818293 150 μL 150µL
2 options
Product Code. 16828293
Brand: Proteintech 113061AP20UL
Rabbit Polyclonal Antibody
Beclin 1 plays a central role in autophagy. It acts as a core subunit of the PI3K complex that mediates formation of phosphatidylinositol 3-phosphate. Different PI3K complex forms are believed to play a role in multiple membrane trafficking pathways - PI3KC3-C1 is involved in the initation of autophagosomes and PI3KC3-C2 is involved in the maturation of autophagosomes and endocytosis. Beclin 1 also plays a role in antiviral host defense.
Specifications
Antigen Beclin 1
Applications Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene BECN1
Gene Accession No. O88597, Q14457, Q91XJ1
Gene Alias ATG6, Beclin 1, beclin 1, autophagy related, beclin1, BECN1, GT197, Protein GT197, VPS30
Gene Symbols Becn1
Host Species Rabbit
Immunogen Beclin 1 Fusion Protein Ag1843
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114558, 56208, 8678
Target Species Human, Rat, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG