Learn More
Chemsplash Beard Mask Non Woven Disposable
Click to view available options
Color:
Blue
Green
Red
White
Yellow
Unit Size:
Pack of 1000
Description
Non-Woven Beard Mask
- Material: Non-Woven
- Features:
- Elasticated
- 100% Latex Free
- 18 in.
- PFAS Free
The Disposable Beard Mask is made from a non-woven material, designed as a one-size-fits-all solution. An elasticated strap ensures a secure fit to the face, making it suitable for use in BRC-approved work areas and the pharmaceutical industry.
Specifications
Specifications
| Type | General Purpose Beard Cover |
| Size | One Size |
| Material | Polypropylene |
| Color | Yellow |
| Disposable | Single-use |
| Latex Free | Yes |
| Length (English) | 18 in. |
| Length (Metric) | 45 cm |