Description
- PP (polypropylene)
- Translucent
- Autoclavable
- Graduated for measuring
Specifications
Specifications
| Color | Translucent |
| Diameter (Metric) Base | 40 mm |
| Diameter (Metric)Top | 50 mm |
| Capacity (Metric) | 100 mL |
| Material | Polypropylene |
| For Use With (Application) | Suitable for Use with Foodstuffs |
| Height (Metric) | 80 mm |
| Includes | Handle |
| Type | Beaker |
| Autoclavable | Autoclavable |
| Show More |