Learn More
Pinnacle Brush™ Beaker Brushes
Product Code. p-8000729
Change view
Click to view available options
Length (Metric):
26 cm
27 cm
36.5 cm
39 cm
Bristle Material:
Fiber
Nylon
Unit Size:
Pack of 10
Product Code. Length (Metric) Bristle Material unitSize
12811934 27 cm Fiber Pack of 10
12398318 26 cm Nylon Pack of 10
12821934 39 cm Fiber Pack of 10
12851934 36.5 cm Nylon Pack of 10
4 options
Product Code. 12811934
Brand: Pinnacle Brush™ T309
Constructed on galvanised steel wire handle. Pinnacle Brush™ Beaker Brushes have nylon bristles for gentle cleaning.
  • Galvanized steel handle
  • Nylon bristles
Specifications
Product Type Bottle Brush
Bristle Material Fiber
Handle Material Metal
Diameter (Metric) 4.5 cm
Length (Metric) 27 cm
Length (Metric) Brush 9.5 cm
Length (Metric) Handle 17.5 cm
Quantity 10 Pk.
Size Brush Circumference: 14 cm