Learn More
Benchmark Scientific™ BeadBath Duo™
Product Code. p-200009132
Change view
Click to view available options
Capacity (Metric):
2 L
Dimensions (D x W x H):
20.3 x 16.5 x 15.2 cm
Dimensions (D x W x H) Interior:
15.2 x 13.9 x 10.1 cm
Unit Size:
Each
Product Code. Capacity (Metric) Dimensions (D x W x H) Dimensions (D x W x H) Interior unitSize
17997070 2 L 20.3 x 16.5 x 15.2 cm 15.2 x 13.9 x 10.1 cm Each
1 options
Product Code. 17997070
Brand: Benchmark Scientific™ B2402E
The BeadBath Duo allows for precise temperature control when using either water or thermal beads.
  • Convert from a water bath to a bead bath at the touch of a button
  • Two separate modes for beads or water ensure optimal performance
  • Quik-Cal™, simple in-lab calibration function
  • Accurate and uniform temperature control
  • No constant refilling
  • Accessory-free securing of containers
  • Ability to run constantly without waiting for heating
Specifications
Capacity (Metric) 2 L
For Use With (Equipment) Tubes, Bottles, Flasks
Temperature Range 5°C to 80°C
Type Bath
Dimensions (D x W x H) 20.3 x 16.5 x 15.2 cm
Dimensions (D x W x H) Interior 15.2 x 13.9 x 10.1 cm
Voltage 230 V
Frequency 50/60 Hz