Learn More
Invitrogen™ Bead Tubes for PureLink™ Microbiome DNA Purification Kit
Product Code. 15659086
Change view
Click to view available options
Quantity:
50 tubes
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15659086 50 tubes Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15659086 Supplier Invitrogen™ Supplier No. A29158
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
These are the same bead tubes that are included in the PureLink™ Microbiome DNA Purification Kit, offered separately for additional applications.
These are the same bead tubes that are included in the PureLink™ Microbiome DNA Purification Kit (Cat. No. A29790), offered separately for additional applications.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at room temperature.
Format Tube
Sample Type Bacteria, Buccal Samples, Cells, Food and Environmental
For Use With (Application) PCR, Real-Time PCR, Sequencing
High-throughput Compatibility Not High-throughput Compatible
Product Line PureLink
Product Type Microbiome DNA Purification Kit
Quantity 50 tubes
Shipping Condition Room Temperature
Target Genomic DNA