missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Vogel and Johnson Agar
Click to view available options
:
Vogel and Johnson Agar (VJ) Agar; 500g
Description
- Formulation: Tryptone 10g/L, Yeast extract 5g/L, Mannitol 10g/L, Dipostassium Phosphate 5g/L, Lithium Chloride 5g/L, Glycine 10g/L, Agar 15g/L, Phenol red 25g/L
Order Info
Prices are for units of one each unless otherwise specified
Certifications
BAM
Specifications
Specifications
| Certifications/Compliance | BAM |
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | Vogel and Johnson Agar |
| Product Type | Agar |
| Formulation | Tryptone 10g/L, Yeast extract 5g/L, Mannitol 10g/L, Dipostassium Phosphate 5g/L, Lithium Chloride 5g/L, Glycine 10g/L, Agar 15g/L, Phenol red 25g/L |
| For Use With (Application) | For early detection of coagulase-positive, mannitol-fermenting staphylococci |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction