Learn More
BD Difco™ Dehydrated Culture Media: Vitamin B12 Assay Medium
Description
Order Info
Prices are for units of one each unless otherwise specified
Certifications
ISO, 9000
Specifications
Specifications
| Certifications/Compliance | ISO, 9000 |
| Content And Storage | 2°C to 8°C |
| Form | Dehydrated |
| Media Type | Vitamin B12 Assay Medium |
| Product Type | Microbiology Media |
| Formulation | Vitamin assay Casamino Acids 12g/L, Dextrose 40g/L, Sodium Acetate 20g/L, L-Cystine 0.2g/L, DL-Tryptophan 0.2g/L, Adenine 20mg/L, Guanine 20mg/L, Uracil 20mg/L, Xanthine 1mg/L, Thiamine Hydrochloride 2mg/L, Riboflavin 2mg/L, Niacin 2mg/L, Calcium Pantothenate 200μg/L, Pyridoxine Hydrochloride 4mg/L, p-Aminobenzoic Acid 200μg/L, Biotin 10μg/L, Folic Acid 100μg/L, Polysorbate 80 2g/L, Dipotassium Phosphate 1g/L, Monopotassium Phosphate 1g/L, Magnesium Sulfate 0.4g/L, Sodium Chloride 20mg/L, Ferrous Sulfate 20mg/L, Manganese Sulfate 20mg/L |
| For Use With (Application) | For determining vitamin B12 concentration by the microbiological assay technique |
| Packaging Type | Bottle |
| Quantity | 100 g |
Safety and Handling
Product Identifier
- vitamin B12 assay medium
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P362+P364-Take off contaminated clothing and wash it before reuse.