Learn More
BD BBL™ Gram Slide
Product Code. 11732964
Change view
Click to view available options
Quantity:
50 Pk.
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11732964 50 Pk. Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11732964 Supplier BD Supplier No. 231401
 Restricted Item
Estimated Shipment: 21-12-2026
Used for evaluating and controlling the quality of Gram stain reagents and techniques. BD Difco™ BBL™ Gram Slide enables easy evaluations.
  • Slide format
  • Used for evaluating: Gram stain reagents 
  • 50 slides
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 30°C
Quantity 50 Pk.