Learn More
BD Propidium Iodide Staining Solution
Product Code. 15818708
Change view
Click to view available options
Quantity:
2 mL
Unit Size:
Each
Product Code. Quantity unitSize
15818708 2 mL Each
1 options
Product Code. 15818708
Brand: BD 556463
Designed for use in two-color Annexin V flow cytometric assays.
  • Detected in the orange range of the spectrum using a 562-588nm band pass filter
  • Used to assess plasma membrane (PM) integrity in Annexin V apoptosis assays
Flow cytometry
TRUSTED_SUSTAINABILITY
Specifications
Type Pharmingen Propidium Iodine Staining Solution
Quantity 2 mL