Learn More
BD Microtainer™ Contact-Activated Lancet
Descripción
These Contact-Activated Lancets are part of the BD Microtainer™ Capillary Products line. This system is designed to provide safety, accuracy and comfort for capillary blood sampling and collection, to meet varying sample requirements and meet the needs of the most fragile patients.
- Has an ergonomic design for a comfortable, secure grip that minimizes slippage during puncture
- Intuitive, one-step procedure enables fast, precise punctures with minimal training
- Activates only when it is positioned and pressed against the skin
- Produces a consistent puncture depth, independent of the clinician’s technique
- Covers only a small area for improved visibility of the site and more accurate positioning
- Design prevents product from being reused, reducing the possibility of contamination
- Automatic retraction of lancet into device minimizes the possibility of injury
- The lot number is laser etched on each lancet for easier tracking
Especificaciones
Especificaciones
| Color | Blue |
| Depth (English) Needle | 0.08 in. |
| Depth (Metric) Needle | 2.0mm |
| Disposable | Single-use |
| Flow Type | High flow |
| For Use With (Application) | Safe for clinician and patient |
| Needle Gauge | 16.5g |
| Quantity | 200 Pack |
| Sterility | Sterile |
| Width (Metric) Blade | 1.5mm |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?