Learn More
BD IMag™ Buffer
Product Code. 15951250
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15951250 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15951250 Supplier BD Supplier No. 552362
 Restricted Item
Estimated Shipment: 12-11-2026
For the optimal labeling of leukocytes using BD IMag particles and for optimal magnetic cell separation using the BD IMagnet™ Cell Separation Magnet
A 10X solution containing bovine serum albumin, EDTA, and sodium azide in phosphate buffered saline (PBS).
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 mL
Content And Storage 4°C
For Use With (Application) Magnetic Cell Separation