Learn More
BD General Use and PrecisionGlide Hypodermic Needles
Product Code. 15892568
Change view
Click to view available options
Gauge:
25 G
Length (English):
1.5 in.
Diameter (English) Outside:
0.02 in.
Thickness (Wall):
Regular
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Gauge Length (English) Diameter (English) Outside Thickness (Wall) Unit Size
15892568 25 G 1.5 in. 0.02 in. Regular Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15892568 Supplier BD Supplier No. 305127
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Fit any Luer-Lok™, slip tip, or eccentric tip syringe
Compatible with any slip tip, Luer-Lok, or eccentric tip syringe.
  • Sheath protects point and cannula
  • Cannula are siliconized to reduce resistance and maintain speed and smoothness throughout injection
  • All have regular bevel, except 14-826-10, which has intradermal bevel
  • 14-826-52, -6A, -6B, and -6C needles have thin-wall design
  • 14-826-6A is for subcutaneous use and has the same flowrate as larger 25-gauge needle
  • 14-826-6B and -6C are for intramuscular use and have the same flowrate as larger 22-gauge needle
  • 14-826-52 has the same flowrate as larger 18-gauge needle
  • Needle hub fits any Luer-Lok or Luer slip syringe tip, reusable or disposable
  • Individual Easy Peel™ blister pack packages boxed in strips of five needles; package carries lot control number Hubs color coded by gauge size: Gray (27-gauge), Tan (26), Blue (25), Turquoise (23), Black (22), Green (21), Yellow (20), Brown (19), Pink (18), and Purple (16)
Order Info
Package carries lot control number to indicate complete sterility.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Conventional Hypodermic Needle
Material Stainless Steel
Sterility Sterile
Cannula Material Siliconized
Color Blue
Connectors Luer-Lok
Diameter (English) Outside 0.02 in.
Diameter (Metric) Outside 0.051 cm
Disposable Single-use
For Use With (Application) Single-use needles minimize cross-infection and provide a factory-sharp point for each injection
For Use With (Equipment) Any Luer-Lok, Slip Tip, or Eccentric Tip Syringe
Gauge 25 G
Hub Material Polypropylene
Includes Protective Plastic Sheath
Latex Free Yes
Length (English) 1.5 in.
Length (Metric) 3.8 cm
Style Bevel Point
Thickness (Wall) Regular