Learn More
BD GasPak™ EZ Gas Generating Systems and Supplies: Incubation Racks
Product Code. 11717204
Change view
Click to view available options
Type:
EZ Incubation Rack
Holds:
Petri Dishes (15 to 18)
Petri Dishes (30 to 33)
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Holds Unit Size
11717204 EZ Incubation Rack Petri Dishes (30 to 33) Each
11707204 EZ Incubation Rack Petri Dishes (15 to 18) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11717204 Supplier BD Supplier No. 260674
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
Catalyst-free convenience for use in producing anaerobic, microaerophilic, or CO2-enriched environments
Catalyst-free convenience for use in producing anaerobic, microaerophilic, or CO2-enriched environments
Holds 30 to 33 petri dishes.
TRUSTED_SUSTAINABILITY
Specifications
Type EZ Incubation Rack
For Use With (Application) Primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria by use of gas generating sachets
Holds Petri Dishes (30 to 33)
Product Line GasPak
Quantity Each
Size Large